2-Chloro-5-methyl-3-nitropyridine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Chloro-3-nitro-5-picoline
A836085-1G
A836085-25G
A836085-10G
103841-188EA
10
USD
103841-188
76845-422
76845-424
77006-768
2-Chloro-5-methyl-3-nitropyridine 98%
2-Chloro-5-methyl-3-nitropyridine
Precaución: for Research Use Only
Formula:
C₆H₅ClN₂O₂ MW: 172.57 Boiling Pt: 290.8 °C (760 mm Hg) Melting Pt: 46…50 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02070020 CAS Number: 23056-40-8 UN: 2923 ADR: 8,6.1 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...