2-Chloro-5-methoxypyridin-3-amine 96%

Supplier: Ambeed
Warning

A193192-100MG A193192-250MG
103816-510EA 101 USD
103816-510 103847-124 76813-986 76813-988
2-Chloro-5-methoxypyridin-3-amine 96%
2-Chloro-5-methoxypyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
Boiling Pt: 311.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11044248
CAS Number: 720666-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR