6-Chloro-m-anisidine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-3-anisidine, 2-Chloro-5-methoxyaniline, 3-Amino-4-chloroanisole

A180458-100G A180458-10G A180458-500G A180458-25G A180458-5G A180458-1G
103807-538EA 6 USD
103807-538 103845-894 76998-650 76998-652 76998-654 77083-386 77083-388 77301-672
6-Chloro-m-anisidine 98%
6-Chloro-m-anisidine

Precaución: for Research Use Only

Formula: C₇H₈ClNO
MW: 157.6 g/mol
Boiling Pt: 135 °C (760 mm Hg)
Melting Pt: 23…27 °C
Storage Temperature: Ambient
MDL Number: MFCD00047830
CAS Number: 2401-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR