2-Chloro-5-isopropoxypyridine 97%
Supplier: Ambeed
|
Warning
|
A149898-250MG
A149898-1G
A149898-5G
103804-098EA
100
USD
103804-098
103842-224
77010-704
77317-738
77317-740
2-Chloro-5-isopropoxypyridine 97%
2-Chloro-5-isopropoxypyridine
Precaución: for Research Use Only
Formula:
C₈H₁₀ClNO MW: 171.624 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19689743 CAS Number: 1204483-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...