2-Chloro-5-iodoanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-4-iodo-2-methoxybenzene

A103230-100MG A103230-250MG A103230-1G A103230-5G
103796-508EA 44 USD
103796-508 77257-338 77257-340 77309-092 77747-768
2-Chloro-5-iodoanisole 98%
2-Chloro-5-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆ClIO
Storage Temperature: Ambient
MDL Number: MFCD00672968
CAS Number: 161949-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR