2-Chloro-5-iodo-4-methoxypyridine 97%

Supplier: Ambeed
Warning

A639485-100MG A639485-250MG
103840-334EA 800 USD
103840-334 77145-250 77747-317
2-Chloro-5-iodo-4-methoxypyridine 97%
2-Chloro-5-iodo-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅ClINO
MW: 269.47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18415711
CAS Number: 1226878-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR