2-Chloro-5-hydroxypyrimidine 98%

Supplier: Ambeed
Warning

A461583-1G A461583-25G A461583-5G A461583-100G A461583-10G A461583-250MG
103827-928EA 9 USD
103827-928 103839-006 76842-888 76842-890 76842-892 76979-552 76979-554 76979-556
2-Chloro-5-hydroxypyrimidine 98%
2-Chloro-5-hydroxypyrimidine

Precaución: For research use only

Formula: C₄H₃ClN₂O
MW: 130.5300 g/mol
Boiling Pt: 328.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09743796
CAS Number: 4983-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR