2-Chloro-5-hydroxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-pyridinol, 6-Chloro-3-pyridinol

A190653-1G A190653-25G A190653-5G A190653-100G A190653-10G A190653-500G
103816-318EA 9 USD
103816-318 103846-918 77084-724 77084-726 77084-728 77274-404 77274-406 77274-408
2-Chloro-5-hydroxypyridine 98%
2-Chloro-5-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄ClNO
MW: 129.55 g/mol
Boiling Pt: 351.9 °C (760 mm Hg)
Melting Pt: 155…158 °C
Flash Pt: 166.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00234050
CAS Number: 41288-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR