(6-Chloro-3-pyridinyl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloropyridine-3-methanol, 2-Chloro-5-hydroxymethylpyridine, 6-Chloro-3-(hydroxymethyl)pyridine

A150715-1G A150715-25G
103804-194EA 5 USD
103804-194 103842-332 77317-860 77317-862
(6-Chloro-3-pyridinyl)methanol 97%
(6-Chloro-3-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Boiling Pt: 276.7 °C (760 mm Hg)
Melting Pt: 51…54 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01075226
CAS Number: 21543-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR