2-Chloro-5-hydroxyisonicotinic acid 97%

Supplier: Ambeed
Warning

A212392-100MG A212392-1G A212392-250MG
103818-196EA 452 USD
103818-196 76975-584 77303-244 77612-086
2-Chloro-5-hydroxyisonicotinic acid 97%
2-Chloro-5-hydroxyisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄ClNO₃
MW: 173.55 g/mol
Boiling Pt: 538.6±50.0 °C (760 mm Hg)
Flash Pt: 279.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13189124
CAS Number: 1060804-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR