2-Chloro-5-hydroxybenzonitrile 97%

Supplier: Ambeed
Warning

A161138-250MG A161138-1G A161138-5G
103805-458EA 69 USD
103805-458 103843-696 76783-860 77300-518 77319-664
2-Chloro-5-hydroxybenzonitrile 97%
2-Chloro-5-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄ClNO
MW: 153.566 g/mol
Boiling Pt: 303.1±27.0 °C (760 mm Hg)
Flash Pt: 137.1 °C
Storage Temperature: Ambient
MDL Number: MFCD09743462
CAS Number: 188774-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR