2-Chloro-5-hydroxybenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A150875-10G
A150875-250MG
A150875-1G
A150875-25G
A150875-5G
103804-210EA
6
USD
103804-210
103842-350
77259-244
77259-246
77299-832
77317-886
77317-888
2-Chloro-5-hydroxybenzoic acid 98%
2-Chloro-5-hydroxybenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₅ClO₃ MW: 172.57 g/mol Boiling Pt: 364.5 °C (760 mm Hg) Melting Pt: 178…180 °C Flash Pt: 174.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038818 CAS Number: 56961-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...