2-Chloro-5-hydrazinylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-hydrazinylpyridine

A178004-250MG A178004-1G
103807-290EA 75 USD
103807-290 103845-630 76813-442 77083-028
2-Chloro-5-hydrazinylpyridine 95%
2-Chloro-5-hydrazinylpyridine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16251142
CAS Number: 145934-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR