2-Chloro-5H-pyrrolo[3,2-d]pyrimidine 97%
Supplier: Ambeed
|
Warning
|
A447194-250MG
A447194-25G
103827-756EA
38
USD
103827-756
103838-824
76789-326
77872-737
2-Chloro-5H-pyrrolo[3,2-d]pyrimidine 97%
2-Chloro-5H-pyrrolo[3,2-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₆H₄ClN₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD11869949 CAS Number: 1119280-66-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...