2-Chloro-5-fluoropyrimidine 98%

Supplier: Ambeed
Warning

A502320-10G A502320-25G A502320-5G A502320-100G
103828-300EA 5 USD
103828-300 103839-390 76818-742 76843-356 76843-358 77594-574
2-Chloro-5-fluoropyrimidine 98%
2-Chloro-5-fluoropyrimidine

Precaución: for Research Use Only

Formula: C₄H₂ClFN₂
MW: 132.52 g/mol
Boiling Pt: 227.4 °C (760 mm Hg)
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788197
CAS Number: 62802-42-0
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR