2-Chloro-5-fluoro-3-hydroxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-fluoropyridin-3-ol

A636980-5G A636980-100MG A636980-10G A636980-250MG A636980-25G
103840-316EA 209 USD
103840-316 76844-488 77157-894 77157-896 77157-898 77157-900
2-Chloro-5-fluoro-3-hydroxypyridine 98%
2-Chloro-5-fluoro-3-hydroxypyridine

Precaución: for Research Use Only

Formula: C₆H₄ClFO
MW: 147.5400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03095268
CAS Number: 884494-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR