2-Chloro-5-fluoronicotinic acid 97%

Supplier: Ambeed
Warning

A262600-100G A262600-10G A262600-25G A262600-5G A262600-1G
103812-570EA 6 USD
103812-570 103852-466 76976-996 76976-998 77092-086 77092-088 77304-850
2-Chloro-5-fluoronicotinic acid 97%
2-Chloro-5-fluoronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₂
MW: 175.5449 g/mol
Boiling Pt: 297 °C (760 mm Hg)
Melting Pt: 141…142 °C
Flash Pt: 133.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03092932
CAS Number: 38186-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR