2-Chloro-5-fluoroisonicotinic acid 98%
Supplier: Ambeed
|
Warning
|
A193095-1G
A193095-25G
A193095-5G
A193095-100MG
A193095-250MG
103816-500EA
14
USD
103816-500
103847-116
77085-014
77085-016
77085-018
77111-844
77111-846
2-Chloro-5-fluoroisonicotinic acid 98%
2-Chloro-5-fluoroisonicotinic acid
Precaución: for Research Use Only
Formula:
C₆H₃ClFNO₂ MW: 175.5500 g/mol Boiling Pt: 383.6 °C (760 mm Hg) Melting Pt: 210…214 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04972381 CAS Number: 884494-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...