2-Chloro-5-fluorobenzotrifluoride 97%

Supplier: Ambeed
Warning

A296433-100G A296433-10G A296433-1G A296433-5G
103814-098EA 6 USD
103814-098 103835-240 76772-528 76772-530 76837-906 76837-908
2-Chloro-5-fluorobenzotrifluoride 97%
2-Chloro-5-fluorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃ClF₄
MW: 198.5500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07782058
CAS Number: 89634-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR