2-Chloro-5-fluorobenzo[d]thiazole 97%

Supplier: Ambeed
Warning

A133815-1G A133815-5G
103801-918EA 65 USD
103801-918 77314-610 77314-612
2-Chloro-5-fluorobenzo[d]thiazole 97%
2-Chloro-5-fluorobenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₇H₃ClFNS
MW: 187.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09749234
CAS Number: 154327-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR