2-Chloro-5-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-fluorobenzeneboronic acid

A211857-1G A211857-25G A211857-5G A211857-100G A211857-10G
103818-144EA 10 USD
103818-144 103848-840 77087-270 77087-272 77087-274 77275-206 77275-208
2-Chloro-5-fluorophenylboronic acid 98%
2-Chloro-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BClFO₂
MW: 174.37 g/mol
Boiling Pt: 309.1 °C (760 mm Hg)
Melting Pt: 78…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06656272
CAS Number: 444666-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR