2-Chloro-5-ethynylpyridine 95%

Supplier: Ambeed
Warning

A110423-100MG A110423-5G
103797-662EA 55 USD
103797-662 103823-468 76809-906 77284-000
2-Chloro-5-ethynylpyridine 95%
2-Chloro-5-ethynylpyridine

Precaución: for Research Use Only

Formula: C₇H₄ClN
MW: 137.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10698136
CAS Number: 263012-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR