2-Chloro-5-ethylpyrimidine 97%

Supplier: Ambeed
Warning

A153296-10G A153296-1G A153296-5G
103804-494EA 13 USD
103804-494 103842-666 77110-394 77318-320 77318-322
2-Chloro-5-ethylpyrimidine 97%
2-Chloro-5-ethylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Boiling Pt: 223 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00799503
CAS Number: 111196-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR