2-Chloro-5-ethoxypyridine 98%

Supplier: Ambeed
Warning

A421831-1G A421831-5G
103827-410EA 118 USD
103827-410 103838-458 76842-074 76842-076
2-Chloro-5-ethoxypyridine 98%
2-Chloro-5-ethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNO
MW: 157.598 g/mol
Boiling Pt: 230.9±20.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09835252
CAS Number: 856851-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR