2-chloro-5,6,7,8-tetrahydroquinoline 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-5,6,7,8-tetrahydroquinoline
A173414-100MG
A173414-250MG
A173414-1G
A173414-5G
103806-816EA
159
USD
103806-816
103845-114
76824-582
76824-584
77082-358
77082-360
2-chloro-5,6,7,8-tetrahydroquinoline 95%
2-chloro-5,6,7,8-tetrahydroquinoline
Precaución: for Research Use Only
Formula:
C₉H₁₀ClN MW: 167.635 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09034992 CAS Number: 21172-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...