2-Chloro-5-(trifluoromethyl)pyridin-4-amine 97%

Supplier: Ambeed
Warning

A127827-25G A127827-1G A127827-5G A127827-250MG
103801-020EA 28 USD
103801-020 76971-940 77313-286 77313-288 77670-814
2-Chloro-5-(trifluoromethyl)pyridin-4-amine 97%
2-Chloro-5-(trifluoromethyl)pyridin-4-amine

Precaución: for Research Use Only

Formula: C₆H₄ClF₃N₂
MW: 196.56 g/mol
Boiling Pt: 287.3±35.0 °C (760 mm Hg)
Flash Pt: 127.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16658126
CAS Number: 1061358-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR