2-Chloro-5-(trifluoromethyl)pyridin-3-amine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-2-chloro-5-(trifluoromethyl)pyridine

A111467-25G A111467-1G A111467-5G A111467-10G
103797-838EA 9 USD
103797-838 103823-658 77025-788 77310-458 77310-460 77570-650
2-Chloro-5-(trifluoromethyl)pyridin-3-amine 98%
2-Chloro-5-(trifluoromethyl)pyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₄ClF₃N₂
MW: 196.557 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07375383
CAS Number: 72587-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR