2-Chloro-5-(trifluoromethyl)nicotinonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-(trifluoromethyl)pyridine-3-carbonitrile

A123462-100MG A123462-250MG A123462-1G
103800-406EA 163 USD
103800-406 77258-152 77258-154 77297-942
2-Chloro-5-(trifluoromethyl)nicotinonitrile 95%
2-Chloro-5-(trifluoromethyl)nicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₂ClF₃N₂
MW: 206.552 g/mol
Boiling Pt: 251.6±35.0 °C (760 mm Hg)
Flash Pt: 105.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10697685
CAS Number: 624734-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR