2-Chloro-5-(trifluoromethyl)nicotinaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-formyl-5-(trifluoromethyl)pyridine, 2-Chloro-5-(trifluoromethyl)pyridine-3-carboxaldehyde

A125066-10G A125066-5G
103800-616EA 99 USD
103800-616 76870-452 77298-030
2-Chloro-5-(trifluoromethyl)nicotinaldehyde 97%
2-Chloro-5-(trifluoromethyl)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO
MW: 209.553 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10696242
CAS Number: 934279-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR