2-Chloro-5-(trifluoromethyl)-4-iodopyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-4-iodo-5-(trifluoromethyl)pyridine
A152784-100G
A152784-10G
A152784-25G
A152784-1G
A152784-5G
103804-432EA
10
USD
103804-432
103842-604
77259-326
77259-328
77299-980
77318-234
77318-236
2-Chloro-5-(trifluoromethyl)-4-iodopyridine 97%
2-Chloro-5-(trifluoromethyl)-4-iodopyridine
Precaución: for Research Use Only
Formula:
C₆H₂ClF₃IN MW: 307.44 g/mol Boiling Pt: 257.6 °C (760 mm Hg) Flash Pt: 110.07 °C Storage Temperature: Ambient |
MDL Number:
MFCD01862099 CAS Number: 505084-55-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...