2-Chloro-5-(methylthio)pyrimidine 95%

Supplier: Ambeed
Warning

A232848-250MG A232848-100MG A232848-5G
103810-858EA 122 USD
103810-858 103850-612 77013-538 77890-351 77890-352
2-Chloro-5-(methylthio)pyrimidine 95%
2-Chloro-5-(methylthio)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂S
Storage Temperature: Refrigerator
MDL Number: MFCD22044944
CAS Number: 115581-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR