2-Hydroxymethyl-5-chloropyrazine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-(hydroxymethyl)pyrazine, (5-Chloropyrazin-2-yl)methanol

A433778-1G A433778-5G A433778-10G A433778-250MG
103827-562EA 15 USD
103827-562 103838-630 76842-336 76842-338 76979-318 76979-320
2-Hydroxymethyl-5-chloropyrazine 95%
2-Hydroxymethyl-5-chloropyrazine

Precaución: For research use only

Formula: C₅H₅ClN₂O
MW: 144.559 g/mol
Boiling Pt: 255.7 °C (760 mm Hg)
Flash Pt: 108.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11110224
CAS Number: 72788-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR