2-Chloro-5-(cyclopropylmethoxy)pyrimidine 97%

Supplier: Ambeed
Warning

A307703-250MG A307703-5G A307703-1G
103814-518EA 152 USD
103814-518 76816-854 76816-856 77305-960
2-Chloro-5-(cyclopropylmethoxy)pyrimidine 97%
2-Chloro-5-(cyclopropylmethoxy)pyrimidine

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O
MW: 184.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20486232
CAS Number: 169677-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR