2-Chloropyrimidine-5-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-chloro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

A155605-1G A155605-5G A155605-10G
103804-782EA 11 USD
103804-782 103842-982 77300-140 77300-142 77592-914
2-Chloropyrimidine-5-boronic acid pinacol ester 97%
2-Chloropyrimidine-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₄BClN₂O₂
MW: 240.494 g/mol
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD11856048
CAS Number: 1003845-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR