2-Chloro-5-pyridineboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-(6-Chloro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Chloropyridine-5-boronic acid Pinacol ester, 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A296629-10G A296629-250MG A296629-5G A296629-1G
103814-108EA 13 USD
103814-108 103835-250 76802-474 76802-476 76837-926 77305-698
2-Chloro-5-pyridineboronic acid pinacol ester 97%
2-Chloro-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅BClNO₂
MW: 239.51
Boiling Pt: 328.3 °C (760 mm Hg)
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD03412793
CAS Number: 444120-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR