2-Chloro-4-nitropyridine 98%

Supplier: Ambeed
Warning

A164836-100G A164836-10G A164836-1G A164836-500G A164836-25G A164836-5G
103844-168EA 12 USD
103844-168 77273-484 77273-486 77273-488 77273-490 77320-360 77320-362
2-Chloro-4-nitropyridine 98%
2-Chloro-4-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₂
MW: 158.54
Boiling Pt: 258.4 °C (760 mm Hg)
Melting Pt: 52…56 °C
Flash Pt: 106.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00661454
CAS Number: 23056-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR