2-Chloro-4-methylpyrimidine-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A194436-100MG A194436-1G
103816-656EA 288 USD
103816-656 103847-266 76769-954 77085-184
2-Chloro-4-methylpyrimidine-5-carboxylic acid 97%
2-Chloro-4-methylpyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
Boiling Pt: 375.3 °C (760 mm Hg)
Flash Pt: 180.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09863167
CAS Number: 188781-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR