5-Amino-2-chloro-4-methylpyrimidine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-methylpyrimidin-5-amine

A262446-100MG A262446-250MG A262446-1G A262446-5G
103812-564EA 35 USD
103812-564 103852-460 76976-992 76976-994 77092-078 77092-080
5-Amino-2-chloro-4-methylpyrimidine 95%
5-Amino-2-chloro-4-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12923128
CAS Number: 20090-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR