2-Chloro-p-toluidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-methylaniline, 4-Amino-3-chlorotoluene

A837938-100G A837938-10G A837938-500G A837938-25G
103841-196EA 7 USD
103841-196 76820-696 76820-698 76820-700 76845-430
2-Chloro-p-toluidine 98%
2-Chloro-p-toluidine

Precaución: for Research Use Only

Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 222.3 °C (760 mm Hg)
Melting Pt: 7 °C
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00007666
CAS Number: 615-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR