2-Chloro-4-methyl-6-(trifluoromethyl)pyrimidine 97%

Supplier: Ambeed
Warning

A100884-100MG A100884-250MG A100884-1G
103795-674EA 75 USD
103795-674 103821-364 76809-228 76809-230 77296-882
2-Chloro-4-methyl-6-(trifluoromethyl)pyrimidine 97%
2-Chloro-4-methyl-6-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄ClF₃N₂
MW: 196.5576 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08690753
CAS Number: 241164-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR