2-Chloro-4-methyl-5-nitropyridine 98%
Supplier: Ambeed
|
Warning
|
A322358-25G
A322358-5G
A322358-100G
A322358-10G
A322358-500G
103836-236EA
6
USD
103836-236
76839-154
76839-156
77141-836
77141-838
77141-840
2-Chloro-4-methyl-5-nitropyridine 98%
2-Chloro-4-methyl-5-nitropyridine
Precaución: for Research Use Only
Formula:
C₆H₅ClN₂O₂ Boiling Pt: 237.1 °C (760 mm Hg) Melting Pt: 37…39 °C Flash Pt: 111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010688 CAS Number: 23056-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...