2-Chloro-4-methyl-3-nitropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-nitro-4-picoline

A183396-25G A183396-5G A183396-100G A183396-10G A183396-500G
103846-200EA 8 USD
103846-200 77083-804 77083-806 77096-930 77096-932 77096-934
2-Chloro-4-methyl-3-nitropyridine 98%
2-Chloro-4-methyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Boiling Pt: 279.6 °C (760 mm Hg)
Melting Pt: 51…53 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00012347
CAS Number: 23056-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR