2-Chloro-4-methoxypyridine 98%

Supplier: Ambeed
Warning

A131283-10G A131283-1G A131283-25G A131283-100G A131283-500G
103801-554EA 5 USD
103801-554 77109-268 77314-042 77314-044 77685-143 77685-144
2-Chloro-4-methoxypyridine 98%
2-Chloro-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆ClNO
Boiling Pt: 213.8 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD02093951
CAS Number: 17228-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR