2-Chloro-4-methoxyphenol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chlor-4-hydroxyanisole

A171762-100G A171762-10G A171762-1G A171762-25G A171762-5G
103844-940EA 9 USD
103844-940 77138-430 77138-432 77138-434 77301-154 77301-156
2-Chloro-4-methoxyphenol 98%
2-Chloro-4-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇ClO₂
MW: 158.58 g/mol
Boiling Pt: 245.3 °C (760 mm Hg)
Melting Pt: 41…44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00070773
CAS Number: 18113-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR