2-Chloro-4-methoxynicotinic acid 98%

Supplier: Ambeed
Warning

A245474-1G A245474-5G A245474-100MG A245474-10G A245474-250MG
103811-648EA 16 USD
103811-648 103851-484 77090-736 77090-738 77165-866 77165-868 77165-870
2-Chloro-4-methoxynicotinic acid 98%
2-Chloro-4-methoxynicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
Boiling Pt: 339.4 °C (760 mm Hg)
Melting Pt: 170…176 °C
Flash Pt: 159 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18378504
CAS Number: 394729-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR