2-Chloro-4-methoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A145442-100G A145442-10G A145442-250MG A145442-1G A145442-25G A145442-5G
103803-534EA 9 USD
103803-534 103841-620 76972-814 76972-816 76972-818 77316-912 77316-914 77316-916
2-Chloro-4-methoxybenzaldehyde 97%
2-Chloro-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
Boiling Pt: 272.5 °C (760 mm Hg)
Melting Pt: 64 °C
Flash Pt: 121.893 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01741722
CAS Number: 54439-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR