2-Chloro-p-anisidine 97%
Supplier: Ambeed
|
Warning
|
A160460-10G
A160460-1G
A160460-25G
A160460-5G
A160460-100G
103805-382EA
6
USD
103805-382
103843-614
76973-568
77300-464
77300-466
77300-468
77881-257
2-Chloro-p-anisidine 97%
2-Chloro-p-anisidine
Precaución: for Research Use Only
Formula:
C₇H₈ClNO Boiling Pt: 284.8 °C (760 mm Hg) Flash Pt: 126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00209639 CAS Number: 29242-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...