2-Chloro-4-methoxy-6-methylpyrimidine 97%

Supplier: Ambeed
Warning

A155810-100MG A155810-250MG A155810-1G
103804-804EA 64 USD
103804-804 103843-006 77259-436 77259-438 77318-746
2-Chloro-4-methoxy-6-methylpyrimidine 97%
2-Chloro-4-methoxy-6-methylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
Boiling Pt: 268.4 °C (760 mm Hg)
Melting Pt: 22…25 °C
Flash Pt: 116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08689881
CAS Number: 22536-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR