2-Chloro-4-methoxy-6-methyl-3-nitropyridine 98%

Supplier: Ambeed
Warning

A109286-250MG A109286-1G
103797-486EA 112 USD
103797-486 103823-280 76982-634 77283-856
2-Chloro-4-methoxy-6-methyl-3-nitropyridine 98%
2-Chloro-4-methoxy-6-methyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₃
MW: 202.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20040427
CAS Number: 179056-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR