2-Chloro-4-iodopyridine 97%

Supplier: Ambeed
Warning

A156369-100G A156369-10G A156369-500G A156369-1G A156369-25G A156369-5G
103804-864EA 9 USD
103804-864 103843-066 77286-274 77286-276 77286-278 77318-830 77318-832 77318-834
2-Chloro-4-iodopyridine 97%
2-Chloro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃ClIN
MW: 239.44 g/mol
Boiling Pt: 255.5 °C (760 mm Hg)
Melting Pt: 42…43 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01861983
CAS Number: 153034-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR